Learn More
Ebro Electronic™ SI 1100 Interface
EBI IF 100 and Winlog.pro software.
Brand: Ebro Electronic™ 1340-6061
This item is not returnable.
View return policy
Description
- With USB connector
- Including antenna AL 111
- With Winlog.pro™ software
- Colored LED signals programming, readout and faulty execution
Order Info
Interface
Specifications
Datalogger System | |
Not Calibrated | |
 IF 100 | |
EBI 10, EBI 100 and EBI 11 data loggers |
No Display | |
None | |
SI 1100 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Ebro Electronic™ SI 1100 Interface > Interface: IF 100