missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ PHPS1 sodium salt

Shp2 (PTPN11) inhibitor

€ 196.00 - € 701.00

Specifications

Molecular Formula C21H14N5O6SNa
Formula Weight 487.42
Product Type Inhibitor
Purity 98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18084892
View Documents
Tocris™
6771/5
5 mg
€ 196.00
5mg
Please to purchase this item. Need a web account? Register with us today!
18706876
View Documents
Tocris™
6771/25
25 mg
€ 701.00
25mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PHPS1 sodium salt is a Shp2 (PTPN11) inhibitor (IC50 = 2.1 μM). Inhibits hepatocyte growth factor-stimulated epithelial cell scattering and branching morphogenesis in vitro. Increases myelin basic protein (MBP) levels in zebrafish embryo spinal cord. Also ameliorates bleomycin-induced pulmonary and dermal fibrosis in vivo.
Specifications

Specifications

C21H14N5O6SNa
487.42
Inhibitor
98%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications