Learn More
Tocris™ PHPS1 sodium salt
Shp2 (PTPN11) inhibitor
Brand: Tocris™ 6771/5
177.95 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
PHPS1 sodium salt is a Shp2 (PTPN11) inhibitor (IC50 = 2.1 μM). Inhibits hepatocyte growth factor-stimulated epithelial cell scattering and branching morphogenesis in vitro. Increases myelin basic protein (MBP) levels in zebrafish embryo spinal cord. Also ameliorates bleomycin-induced pulmonary and dermal fibrosis in vivo.Specifications
5 mg | |
487.42 | |
98% |
C21H14N5O6SNa | |
Inhibitor |