Learn More
Tocris™ PHPS1 sodium salt
Shp2 (PTPN11) inhibitor
Brand:  Tocris™ 6771/5
177.95 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18084892
  • € 196.00 / 5mg
Explore more special offers
Description
Description
PHPS1 sodium salt is a Shp2 (PTPN11) inhibitor (IC50 = 2.1 μM). Inhibits hepatocyte growth factor-stimulated epithelial cell scattering and branching morphogenesis in vitro. Increases myelin basic protein (MBP) levels in zebrafish embryo spinal cord. Also ameliorates bleomycin-induced pulmonary and dermal fibrosis in vivo.
Specifications
Specifications
5 mg
487.42
98%
C21H14N5O6SNa
Inhibitor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers