Learn More
Thermo Scientific™ Orion™ Automatic Stirrer Probe and Paddle
Use this stirrer probe with our benchtop meters to stir solutions for better accuracy without the difficulty of using a stir plate and bar.
Brand:  Thermo Scientific™ 096021
 View more versions of this product
Product Code. 11777757
  • € 68.20 / Pack of 3
In Stock
to see stock.
Explore more special offers
Description
Use the Thermo Scientific™ Orion Star™ Stirrer Probe to stir standards and samples for better measurement accuracy without the difficulty of separate stir plate and bars. Product is intended for use with Thermo Scientific™ Orion™ Versa Star meters, Thermo Scientific™ Orion Star™ A210 series benchtop meters and legacy Orion Star™ series benchtop meters. The Orion Star™ Stirrer Probe is powered and controlled by the meter with five optimized stirring speeds and fits in the electrode stand for easy movement in and out samples and simplified rinsing. Paddles can be replaced to extend the life of Orion Star™ Stirrer Probes.
Fits Swing Arm Electrode Holder 96019.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.6 in. round (stirring paddle)
1.5 cm round (stirring paddle)
1.2 x 1.5 x 1.5 cm (0.5 x 0.6 x 0.6 in.)
Orion Star Series Automatic Stirrer Probe and Paddle
Orion Star series stirrer replacement paddles, 3 pack
Meter Accessories
Three replaceable stirring paddles for the Orion Star series stirrer probe
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Orion™ Automatic Stirrer Probe and Paddle >
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.