missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(3-nitrobenzyl)amine, 97+%, Thermo Scientific™

€ 33.90 - € 127.00

Chemical Identifiers

CAS 19499-61-7
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
InChI Key NTPAPKLGADEFAM-UHFFFAOYSA-N
Synonym n-methyl-1-3-nitrophenyl methanamine, n-methyl-n-3-nitrobenzyl amine, methyl 3-nitrophenyl methyl amine, benzenemethanamine, n-methyl-3-nitro, 3-nitro-n-methylbenzylamine, n-methyl-3-nitrobenzylamine, n-3-nitrophenylmethyl methylamine, n-methyl-1-3-nitrophenyl methylamine
PubChem CID 409439
SMILES CNCC1=CC(=CC=C1)[N+](=O)[O-]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10483353
View Documents
Thermo Scientific Maybridge
MO00890DA
1 g Amber Glass Bottle
€ 33.90
1g
Please to purchase this item. Need a web account? Register with us today!
10123674
View Documents
Thermo Scientific Maybridge
MO00890DE
5 g Amber Glass Bottle
€ 127.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

19499-61-7
166.18
n-methyl-1-3-nitrophenyl methanamine, n-methyl-n-3-nitrobenzyl amine, methyl 3-nitrophenyl methyl amine, benzenemethanamine, n-methyl-3-nitro, 3-nitro-n-methylbenzylamine, n-methyl-3-nitrobenzylamine, n-3-nitrophenylmethyl methylamine, n-methyl-1-3-nitrophenyl methylamine
CNCC1=CC(=CC=C1)[N+](=O)[O-]
C8H10N2O2
NTPAPKLGADEFAM-UHFFFAOYSA-N
409439

Specifications

19499-61-7
C8H10N2O2
NTPAPKLGADEFAM-UHFFFAOYSA-N
166.18
166.18
N-Methyl-N-(3-nitrobenzyl)amine
95%
n-methyl-1-3-nitrophenyl methanamine, n-methyl-n-3-nitrobenzyl amine, methyl 3-nitrophenyl methyl amine, benzenemethanamine, n-methyl-3-nitro, 3-nitro-n-methylbenzylamine, n-methyl-3-nitrobenzylamine, n-3-nitrophenylmethyl methylamine, n-methyl-1-3-nitrophenyl methylamine
CNCC1=CC(=CC=C1)[N+](=O)[O-]
409439
95%
Videos
SDS
Documents

Documents

Product Certifications