missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(3-nitrobenzyl)amine, 97+%, Thermo Scientific™

Product Code. 10123674
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 19499-61-7
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
InChI Key NTPAPKLGADEFAM-UHFFFAOYSA-N
Synonym n-methyl-1-3-nitrophenyl methanamine, n-methyl-n-3-nitrobenzyl amine, methyl 3-nitrophenyl methyl amine, benzenemethanamine, n-methyl-3-nitro, 3-nitro-n-methylbenzylamine, n-methyl-3-nitrobenzylamine, n-3-nitrophenylmethyl methylamine, n-methyl-1-3-nitrophenyl methylamine
PubChem CID 409439
SMILES CNCC1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 166.18
Percent Purity 95%
Chemical Name or Material N-Methyl-N-(3-nitrobenzyl)amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'