missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ MAS™ CardioImmune™ XL Controls
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls.
€ 341.00
Specifications
Content And Storage | 3 year shelf life at -25° to -15°C; 30 days open-vial at 2° to 8°C (for hsCRP, Digitoxin, Troponin I and Homocysteine); 15 days open-vial at 2° to 8°C (for BNP32, Myoglobin (when a dropper tip is used), proBNP, Troponin T, CK-MB) |
---|---|
Detectable Analytes | Brain Natriuretic Peptide-32 (BNP-32), Myoglobin, Creatinine Kinase-MB (CK-MB), N-Terminal Brain Natriuretic Peptide (NT-proBNP), Digitoxin, Troponin I, Homocysteine, Troponin T, hcCRP |
Sample Type | Serum |
Type | Control |
Packaging | 6 Pk. |
Product Code | Brand | Description | Level | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Level | Price | Quantity & Availability | ||||
15160027
|
Thermo Scientific™
CAI-XLL |
CardioImmune XL Control, Assayed | Level L |
€ 341.00
18mL |
Estimated Shipment: 17-02-2025
Log in to see stock. |
||||
15170027
|
Thermo Scientific™
CAI-XL1 |
CardioImmune XL Control, Assayed | 1 |
€ 341.00
18mL |
Estimated Shipment: 17-02-2025
Log in to see stock. |
||||
15180027
|
Thermo Scientific™
CAI-XL2 |
CardioImmune XL Control, Assayed | 2 |
€ 341.00
18mL |
Estimated Shipment: 17-02-2025
Log in to see stock. |
||||
15190027
|
Thermo Scientific™
CAI-XL3 |
CardioImmune XL Control, Assayed | 3 |
€ 341.00
18mL |
Estimated Shipment: 17-02-2025
Log in to see stock. |
||||
Description
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls. These controls are also available in a convenient Vista™ vial format.
- Human serum matrix
- Offers 4 distinctive levels
- Assayed values available for most major analyzers
- Available in a tri-level multipack
Specifications
3 year shelf life at -25° to -15°C; 30 days open-vial at 2° to 8°C (for hsCRP, Digitoxin, Troponin I and Homocysteine); 15 days open-vial at 2° to 8°C (for BNP32, Myoglobin (when a dropper tip is used), proBNP, Troponin T, CK-MB) | |
Serum | |
6 Pk. |
Brain Natriuretic Peptide-32 (BNP-32), Myoglobin, Creatinine Kinase-MB (CK-MB), N-Terminal Brain Natriuretic Peptide (NT-proBNP), Digitoxin, Troponin I, Homocysteine, Troponin T, hcCRP | |
Control |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title