Learn More
Thermo Scientific™ MAS™ CardioImmune™ XL Controls
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls.
Brand:  Thermo Scientific™ CAI-XL1
 View more versions of this product
Product Code. 15170027
  • € 341.00 / 18mL
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls. These controls are also available in a convenient Vista™ vial format.
  • Human serum matrix
  • Offers 4 distinctive levels
  • Assayed values available for most major analyzers
  • Available in a tri-level multipack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3 year shelf life at -25° to -15°C; 30 days open-vial at 2° to 8°C (for hsCRP, Digitoxin, Troponin I and Homocysteine); 15 days open-vial at 2° to 8°C (for BNP32, Myoglobin (when a dropper tip is used), proBNP, Troponin T, CK-MB)
Brain Natriuretic Peptide-32 (BNP-32), Myoglobin, Creatinine Kinase-MB (CK-MB), N-Terminal Brain Natriuretic Peptide (NT-proBNP), Digitoxin, Troponin I, Homocysteine, Troponin T, hcCRP
Control
6 Pk.
CardioImmune XL Control, Assayed
Serum
1
3 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers