Learn More
Thermo Scientific™ MAS™ CardioImmune™ XL Controls
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls.
Brand: Thermo Scientific™ CAI-XL2
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls. These controls are also available in a convenient Vista™ vial format.
- Human serum matrix
- Offers 4 distinctive levels
- Assayed values available for most major analyzers
- Available in a tri-level multipack
Especificaciones
3 year shelf life at -25° to -15°C; 30 days open-vial at 2° to 8°C (for hsCRP, Digitoxin, Troponin I and Homocysteine); 15 days open-vial at 2° to 8°C (for BNP32, Myoglobin (when a dropper tip is used), proBNP, Troponin T, CK-MB) | |
Brain Natriuretic Peptide-32 (BNP-32), Myoglobin, Creatinine Kinase-MB (CK-MB), N-Terminal Brain Natriuretic Peptide (NT-proBNP), Digitoxin, Troponin I, Homocysteine, Troponin T, hcCRP | |
Control | |
6 Pk. |
CardioImmune XL Control, Assayed | |
Serum | |
2 | |
3 mL |