Learn More
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
Brand: Poeppelmann™ 61000330000
Description
- Universal use as a plug or cap
- Acc. with VDA Band 19 and ISO 16232
- Tapered design ideal for accomodating tolerances
Specifications
Yellow | |
Used universally as plug or cap, Tapered design is ideal for accommHodating tolerances | |
46.1 mm | |
42.8 dia. x 19mmH | |
Closing Plug |
44.4 dia. x 19mmH | |
51 mm | |
100 Pack | |
Polyethylene |