Learn More
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
Brand: Poeppelmann™ 61000160000
Description
- Universal use as a plug or cap
- Acc. with VDA Band 19 and ISO 16232
- Tapered design ideal for accomodating tolerances
Specifications
Yellow | |
Used universally as plug or cap, Tapered design is ideal for accommodating tolerances | |
100 Pack | |
Polyethylene |
17.5 mm | |
15 mm | |
24 mm | |
Closing Plug |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs > Interior Dimensions: 17.5 dia. x 15mmH; Exterior Dimensions: 24 dia. x 15mmH