Learn More
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
Brand:  Poeppelmann™ 61000270000
 View more versions of this product
Product Code. 10043384
  • € 47.68 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Universal use as a plug or cap
  • Acc. with VDA Band 19 and ISO 16232
  • Tapered design ideal for accomodating tolerances
Specifications
Specifications
Yellow
Used universally as plug or cap, Tapered design is ideal for accommHodating tolerances
33.4 mm
39.7 dia. x 17mmH
Closing Plug
31.4 dia. x 31mmH
38 mm
100 Pack
Polyethylene
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs > Interior Dimensions: 31.4 dia. x 31mmH; Exterior Dimensions: 39.7 dia. x 17mmH