Learn More
Irinotecan hydrochloride, MedChemExpress
MedChemExpress Irinotecan hydrochloride ((+)-Irinotecan hydrochloride) is a topoisomerase I inhibitor mainly used to treat colon cancer and rectal cancer.
€ 120.15 - € 523.80
Specifications
Chemical Name or Material | Irinotecan hydrochloride |
---|---|
CAS | 100286-90-6 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C33H39ClN4O6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16487116
|
Medchem Express
HY-16562A |
100 mg |
€ 196.50
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16483117
|
Medchem Express
HY-16562A |
50 mg |
€ 120.15
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16431226
|
Medchem Express
HY-16562A |
200 mg |
€ 279.45
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16401816
|
Medchem Express
HY-16562A |
500 mg |
€ 523.80
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Irinotecan hydrochloride ((+)-Irinotecan hydrochloride) is a topoisomerase I inhibitor mainly used to treat colon cancer and rectal cancer.Specifications
Irinotecan hydrochloride | |
White | |
C33H39ClN4O6 | |
DMSO : 125 mg/mL (200.60 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (5.34 mM; Need ultrasonic) | |
623.14 | |
98.03% | |
Powder |
100286-90-6 | |
Research | |
(+)-Irinotecan hydrochlorideCPT-11 hydrochloride | |
O=C(N1CCC(N2CCCCC2)CC1)OC3=CC=C4N=C5C(CN6C(C(COC([C@@]7(CC)O)=O)=C7C=C65)=O)=C(CC)C4=C3.[H]Cl | |
623.14 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
H302
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.