Learn More
Irinotecan hydrochloride, MedChemExpress
MedChemExpress Irinotecan hydrochloride ((+)-Irinotecan hydrochloride) is a topoisomerase I inhibitor mainly used to treat colon cancer and rectal cancer.
Brand: Medchem Express HY-16562A
This item is not returnable.
View return policy
Description
Irinotecan hydrochloride ((+)-Irinotecan hydrochloride) is a topoisomerase I inhibitor mainly used to treat colon cancer and rectal cancer.Specifications
Irinotecan hydrochloride | |
White | |
C33H39ClN4O6 | |
(+)-Irinotecan hydrochlorideCPT-11 hydrochloride | |
O=C(N1CCC(N2CCCCC2)CC1)OC3=CC=C4N=C5C(CN6C(C(COC([C@@]7(CC)O)=O)=C7C=C65)=O)=C(CC)C4=C3.[H]Cl | |
623.14 | |
Research | |
Cancer-programmed cell death |
100286-90-6 | |
Research | |
50 mg | |
DMSO : 125 mg/mL (200.60 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (5.34 mM; Need ultrasonic) | |
623.14 | |
98.03% | |
Powder |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only