Learn More
Keystone Cleanroom Products Heavy Duty SMS Autoclave Bag
This product's superior barrier prevents liquid and microbial penetration.
€ 1315.00 - € 2720.00
Specifications
Product Type | Autoclave Bag |
---|---|
Basis Weight | 50 gsm |
Autoclavable | Autoclavable |
Certifications/Compliance | CoC |
Closure Type | Self-adhesive |
Product Code | Brand | Capacity (English) | Capacity (Metric) | Length (Metric) | Length (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Capacity (Metric) | Length (Metric) | Length (English) | Price | Quantity & Availability | |||
17852477
![]() |
Keystone Cleanroom Products
BGSMS3SS09.516 |
135 oz. | 3.99 L | 41 cm | 16 in. |
€ 1315.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17862477
![]() |
Keystone Cleanroom Products
BGSMS3SS1217 |
176 oz. | 5.2 L | 43.2 cm | 17 in. |
€ 1405.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17892477
![]() |
Keystone Cleanroom Products
BGSMS3SS1221 |
220 oz. | 6.51 L | 53.3 cm | 21 in. |
€ 1500.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17872477
![]() |
Keystone Cleanroom Products
BGSMS3SS1318 |
198 oz. | 5.86 L | 45.7 cm | 18 in. |
€ 1500.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17882477
![]() |
Keystone Cleanroom Products
BGSMS3SS14.522 |
312 oz. | 8.46 L | 55.9 cm | 22 in. |
€ 1570.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17802487
![]() |
Keystone Cleanroom Products
BGSMS3SS1520 |
258 oz. | 7.6 L | 50.8 cm | 20 in. |
€ 1570.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17812487
![]() |
Keystone Cleanroom Products
BGSMS3SS1622 |
330 oz. | 9.75 L | 55.9 cm | 22 in. |
€ 1595.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17832487
![]() |
Keystone Cleanroom Products
BGSMS3SS1824 |
408 oz. | 12 L | 61 cm | 24 in. |
€ 1615.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17822487
![]() |
Keystone Cleanroom Products
BGSMS3SS1830 |
510 oz. | 15 L | 76.2 cm | 30 in. |
€ 1640.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17862487
![]() |
Keystone Cleanroom Products
BGSMS3SS2028 |
550 oz. | 16.3 L | 71 cm | 28 in. |
€ 1640.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17842487
![]() |
Keystone Cleanroom Products
BGSMS3SS2126 |
486 oz. | 14.4 L | 66 cm | 26 in. |
€ 1640.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17852487
![]() |
Keystone Cleanroom Products
BGSMS3SS2424 |
510 oz. | 15 L | 61 cm | 24 in. |
€ 1735.00
Pack of 125 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17872487
![]() |
Keystone Cleanroom Products
BGSMS3SS2430 |
662 oz. | 19.6 L | 76.2 cm | 30 in. |
€ 1875.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17882487
![]() |
Keystone Cleanroom Products
BGSMS3SS2435 |
790 oz. | 23.4 L | 88.9 cm | 35 in. |
€ 2435.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17892467
![]() |
Keystone Cleanroom Products
BGSMS3SS2630 |
705 oz. | 20.8 L | 76.2 cm | 30 in. |
€ 1900.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17802497
![]() |
Keystone Cleanroom Products
BGSMS3SS2836 |
920 oz. | 27.2 L | 91.4 cm | 36 in. |
€ 2530.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17822477
![]() |
Keystone Cleanroom Products
BGSMS3SS3030 |
870 oz. | 25.7 L | 76.2 cm | 30 in. |
€ 2435.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17812477
![]() |
Keystone Cleanroom Products
BGSMS3SS3636 |
1260 oz. | 37.3 L | 91.4 cm | 36 in. |
€ 2720.00
Pack of 250 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17842477
![]() |
Keystone Cleanroom Products
BGSMS3SS3648 |
1680 oz. | 49.7 L | 121.9 cm | 48 in. |
€ 1640.00
Pack of 125 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
17832477
![]() |
Keystone Cleanroom Products
BGSMS3SS4848 |
2256 oz. | 66.7 L | 121.9 cm | 48 in. |
€ 1875.00
Pack of 125 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
Description
- Convenient, time saving self-seal closure system
- Significant improvement versus cutting and wrapping with sheets and tape
- Manufactured and packaged in a certified ISO6 Cleanroom
- Strong, rip and tear resistant 50 gsm SMS material
- Sub packed into sealed labeled poly bags with manufacturing lot number and date of manufacture for traceability
- Made in the USA
Specifications
Autoclave Bag | |
Autoclavable | |
Self-adhesive | |
Single-use | |
Non-woven Fabric | |
50 gsm | |
50 gsm |
50 gsm | |
CoC | |
Light Blue | |
Steam Sterilisation | |
Non-sterile | |
Yes |
Product Title
Keystone Cleanroom Products Heavy Duty SMS Autoclave Bag