Learn More
Keystone Cleanroom Products Heavy Duty SMS Autoclave Bag
GREENER_CHOICE
This product's superior barrier prevents liquid and microbial penetration.
€ 1315.00 - € 2720.00
Specifications
Product Type Autoclave Bag
Basis Weight 50 gsm
Autoclavable Autoclavable
Certifications/Compliance CoC
Closure Type Self-adhesive
View More Specs
Products 20
Product Code Brand Capacity (English) Capacity (Metric) Length (Metric) Length (English) Price Quantity & Availability  
Product Code Brand Capacity (English) Capacity (Metric) Length (Metric) Length (English) Price Quantity & Availability  
17852477
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS09.516
135 oz. 3.99 L 41 cm 16 in.
€ 1315.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17862477
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS1217
176 oz. 5.2 L 43.2 cm 17 in.
€ 1405.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17892477
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS1221
220 oz. 6.51 L 53.3 cm 21 in.
€ 1500.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17872477
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS1318
198 oz. 5.86 L 45.7 cm 18 in.
€ 1500.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17882477
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS14.522
312 oz. 8.46 L 55.9 cm 22 in.
€ 1570.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17802487
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS1520
258 oz. 7.6 L 50.8 cm 20 in.
€ 1570.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17812487
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS1622
330 oz. 9.75 L 55.9 cm 22 in.
€ 1595.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17832487
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS1824
408 oz. 12 L 61 cm 24 in.
€ 1615.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17822487
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS1830
510 oz. 15 L 76.2 cm 30 in.
€ 1640.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17862487
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS2028
550 oz. 16.3 L 71 cm 28 in.
€ 1640.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17842487
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS2126
486 oz. 14.4 L 66 cm 26 in.
€ 1640.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17852487
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS2424
510 oz. 15 L 61 cm 24 in.
€ 1735.00
Pack of 125
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17872487
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS2430
662 oz. 19.6 L 76.2 cm 30 in.
€ 1875.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17882487
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS2435
790 oz. 23.4 L 88.9 cm 35 in.
€ 2435.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17892467
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS2630
705 oz. 20.8 L 76.2 cm 30 in.
€ 1900.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17802497
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS2836
920 oz. 27.2 L 91.4 cm 36 in.
€ 2530.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17822477
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS3030
870 oz. 25.7 L 76.2 cm 30 in.
€ 2435.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17812477
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS3636
1260 oz. 37.3 L 91.4 cm 36 in.
€ 2720.00
Pack of 250
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17842477
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS3648
1680 oz. 49.7 L 121.9 cm 48 in.
€ 1640.00
Pack of 125
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
17832477
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGSMS3SS4848
2256 oz. 66.7 L 121.9 cm 48 in.
€ 1875.00
Pack of 125
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
Description
Description
  • Convenient, time saving self-seal closure system
  • Significant improvement versus cutting and wrapping with sheets and tape
  • Manufactured and packaged in a certified ISO6 Cleanroom
  • Strong, rip and tear resistant 50 gsm SMS material
  • Sub packed into sealed labeled poly bags with manufacturing lot number and date of manufacture for traceability
  • Made in the USA
Specifications
Specifications
Autoclave Bag
Autoclavable
Self-adhesive
Single-use
Non-woven Fabric
50 gsm
50 gsm
50 gsm
CoC
Light Blue
Steam Sterilisation
Non-sterile
Yes
Documents
Product Certifications
Product Content Correction
Product Title
Keystone Cleanroom Products Heavy Duty SMS Autoclave Bag