Learn More
Eppendorf™ Galaxy™ 48 R Incubators
When a high level of cell culture isolation is required or very limited space is available in the lab, the stackable Galaxy 48R is the right choice. The high temperature disinfection, a seamless chamber and a fanless design makes it easy to clean and provides high contamination protection.
Brand:  Eppendorf™ CO48312061
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 15783395
Description
Description
  • Small footprint: can be used in smaller laboratories.
  • Large backlit display with on screen help menu.
  • Control and data logging of temperature, CO2 , door openings, and alarms.
  • Fanless design offers more usable space.
  • Six-sided direct heating system provides a uniform incubated environment.
  • Deep-drawn, easy-to-clean stainless steel chamber without seams or welds.
  • Low gas consumption.
  • Integrated alarm relay.
  • Available options based on model number:
    • O2 gas control.
    • Humidity package.
    • High temperature disinfection (4 hour at 120°C).
    • O2 control (e.g., for hypoxic studies).
    • 2-split inner glass door.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
International Power Supply, High Temperature Disinfection, Humidity Package
Stainless Steel
0.1 to 19%
CO2 Incubator
48 L
IR Sensor
0.1 to 19% O2 Control
2-Split Inner Door
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Galaxy™ 48 R Incubators > 2-Split Inner Door, International Power Supply, High Temperature Disinfection, Humidity Package, 0.1-19% O2 Control