Learn More
Buerkle™ Stainless Steel Conical QualiRod
Conical hand-drill, conically shaped, with sharp cutting edges and a solid handle. Ideal for cutting out samples for quality control from soft, paste-like substances (cheese, butter, clay, soap, wax, etc.)
Brand:  Buerkle™ 5311-0001
 View more versions of this product
Product Code. 10690543
  • € 89.90 / Each
In Stock
to see stock.
Explore more special offers
Description
A conically shaped hand-drill, with sharp cutting edges and a solid handle. Ideal for cutting out samples for quality control from soft, paste-like substances (cheese, butter, clay, soap, wax, etc.).
  • Stainless steel V2A (1.4301)
  • Sterilizable
  • According to EN/ISO 707
Specifications
Specifications
Stainless Steel (AISI 304)
Sterilisable conical QualiRod
Hand Drilling Conical Shaped Samples for Quality Control Checks From Solft Paste-Like Substances Such as Cheese, Butter, Clay, Soat, Wax Etc.
Autoclavable
Sterilisable Conical QualiRod
110 mm
Videos
  • Buerkle - Zone samplers - Multi sampler, All-layer sampler, Uno sampler
    "Bürkle zone samplers are used for taking samples of bulk goods of any kind. The big advantage of zone samplers is that all-layer samples can be collected from all layers of the container."
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Buerkle Zone samplers Multi sampler, All-layer sampler, Uno sampler
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Buerkle™ Stainless Steel Conical QualiRod > Drilling Depth: 110mm