Learn More
OHAUS™ Cup Head
OHAUS™ Cup Head is an accessory for use with a vortex mixer.
€ 37.80 - € 41.85
Specifications
Product Type | Cup Head |
---|---|
Material | Rubber |
For Use With (Equipment) | Vortex Mixers |
Packaging Quantity | 1 Ea. |
Description
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Cup Head | |
Rubber | |
Vortex Mixers | |
1 Ea. |