Learn More
OHAUS™ Cup Head
OHAUS™ Cup Head is an accessory for use with a vortex mixer.
Brand: OHAUS™ 30400210
37.33 EUR valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Cup Head | |
Gray | |
Vortex Mixers |
Rubber | |
Cup Head, VXHD and VXMP Vortex Mixers | |
1 Ea. |