Learn More
OHAUS™ Cup Head
OHAUS™ Cup Head is an accessory for use with a vortex mixer.
Brand: OHAUS™ 30400235
Additional Details : Weight : 0.01000kg
Description
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Cup Head | |
Black | |
Vortex Mixers |
Rubber | |
Cup Head | |
1 Ea. |