Learn More
Tenak™ Comfort Rack for Microtiter Plates for Upright Freezers, 45mm Shelf Height
Providing a complete range of racks for microtiter plates in chest freezers. Tenak™ Comfort Racks for Microtiter Plates in Upright Freezers are stackable.
€ 179.00 - € 343.00
Specifications
Product Type | Microtiter Plate Rack |
---|---|
Stackable | Yes |
For Use With (Equipment) | Microtiter Plates, Upright Freezers |
No. per Pack | 1 Pack |
Width (Metric) | 135 mm |
Product Code | Brand | Dimensions (L x W x H) | Array | Holds | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Array | Holds | Price | Quantity & Availability | |||
12407040
|
Tenak™
TE35420 |
360 x 135 x 140mm | 3 x 4 | Plates (12) |
€ 179.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12407060
|
Tenak™
TE35480 |
450 x 135 x 140mm | 3 x 5 | Plates (15) |
€ 197.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12417040
|
Tenak™
TE35422 |
360 x 135 x 186mm | 4 x 4 | Plates (16) |
€ 203.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12417060
|
Tenak™
TE35482 |
450 x 135 x 186mm | 4 x 5 | Plates (20) |
€ 215.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12427040
|
Tenak™
TE35424 |
360 x 135 x 233mm | 5 x 4 | Plates (20) |
€ 226.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12427060
|
Tenak™
TE35484 |
450 x 135 x 233mm | 5 x 5 | Plates (25) |
€ 240.00
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||
12427080
|
Tenak™
TE35540 |
540 x 135 x 140mm | 3 x 6 | Plates (18) |
€ 210.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12437040
|
Tenak™
TE35426 |
360 x 135 x 279mm | 6 x 4 | Plates (24) |
€ 250.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12437060
|
Tenak™
TE35486 |
450 x 135 x 279mm | 6 x 5 | Plates (30) |
€ 263.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12437080
|
Tenak™
TE35542 |
540 x 135 x 186mm | 4 x 6 | Plates (24) |
€ 238.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12437100
|
Tenak™
TE35590 |
630 x 135 x 140mm | 3 x 7 | Plates (21) |
€ 229.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12447060
|
Tenak™
TE35488 |
450 x 135 x 325mm | 7 x 5 | Plates (35) |
€ 286.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12447080
|
Tenak™
TE35544 |
540 x 135 x 233mm | 5 x 6 | Plates (30) |
€ 267.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12447100
|
Tenak™
TE35592 |
630 x 135 x 186mm | 4 x 7 | Plates (28) |
€ 257.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12457060
|
Tenak™
TE35490 |
450 x 135 x 372mm | 8 x 5 | Plates (40) |
€ 305.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12457080
|
Tenak™
TE35546 |
540 x 135 x 279mm | 6 x 6 | Plates (36) |
€ 295.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12457100
|
Tenak™
TE35594 |
630 x 135 x 233mm | 5 x 7 | Plates (35) |
€ 286.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12467080
|
Tenak™
TE35548 |
540 x 135 x 325mm | 7 x 6 | Plates (42) |
€ 324.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
12467100
|
Tenak™
TE35596 |
630 x 135 x 279mm | 6 x 7 | Plates (42) |
€ 314.00
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||
12477100
|
Tenak™
TE35598 |
630 x 135 x 325mm | 7 x 7 | Plates (49) |
€ 343.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||
Description
- Racks can either be used as one plate per shelf or the plates can be stacked
- With an alert preventing the shelves from sliding out
- External footprint is 135 x 92mm
Specifications
Microtiter Plate Rack | |
Microtiter Plates, Upright Freezers | |
135 mm |
Yes | |
1 Pack |