Learn More
Tenak™ Comfort Rack for Microtiter Plates for Upright Freezers, 45mm Shelf Height
Providing a complete range of racks for microtiter plates in chest freezers. Tenak™ Comfort Racks for Microtiter Plates in Upright Freezers are stackable.
Brand:  Tenak™ TE35488
Additional Details : Weight : 0.55000kg
 View more versions of this product
Product Code. 12447060
  • € 286.00 / Each
Estimated Shipment: 14-10-2024
to see stock.
Add to basket
Description
Description
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • With an alert preventing the shelves from sliding out
  • External footprint is 135 x 92mm
Specifications
Specifications
Microtiter Plate Rack
Plates (35)
Yes
1 Pack
135 mm
450 mm
7 x 5
35
Microtiter Plates, Upright Freezers
450 x 135 x 325mm
325 mm