missing translation for 'onlineSavingsMsg'
Learn More

CNX-2006, MedChemExpress

MedChemExpress CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.

€ 143.10 - € 1548.00

Specifications

Chemical Name or Material CNX-2006
CAS 1375465-09-0
Color Gray
Purity Grade Notes Research
Molecular Formula C26H27F4N7O2
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16475898
Encompass
View Documents
Medchem Express
HY-13897
10 mg
€ 292.50
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16462047
Encompass_Preferred
View Documents
Medchem Express
HY-13897
50 mg
€ 778.95
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16458537
Encompass_Preferred
View Documents
Medchem Express
HY-13897
5 mg
€ 143.10
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16406356
Encompass
View Documents
Medchem Express
HY-13897
100 mg
€ 1548.00
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.
Specifications

Specifications

CNX-2006
Gray
C26H27F4N7O2
C=CC(NC1=CC=CC(NC2=NC(NC3=CC=C(NC4CN(CCF)C4)C=C3OC)=NC=C2C(F)(F)F)=C1)=O
545.53
Research
Cancer-Kinase/protease
1375465-09-0
Research
DMSO : ≥ 52 mg/mL (95.32 mM)
545.53
99.68%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.