Learn More
CNX-2006, MedChemExpress
MedChemExpress CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.
Brand: Medchem Express HY-13897
This item is not returnable.
View return policy
Description
CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.Specifications
CNX-2006 | |
Gray | |
C26H27F4N7O2 | |
DMSO : ≥ 52 mg/mL (95.32 mM) | |
545.53 | |
99.68% | |
Solid |
1375465-09-0 | |
Research | |
100 mg | |
C=CC(NC1=CC=CC(NC2=NC(NC3=CC=C(NC4CN(CCF)C4)C=C3OC)=NC=C2C(F)(F)F)=C1)=O | |
545.53 | |
Research | |
Cancer-Kinase/protease |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only