Learn More
CNX-2006, MedChemExpress
MedChemExpress CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.
Brand:  Medchem Express HY-13897
Encompass_Preferred
 View more versions of this product
Product Code. 16462047
  • € 778.95 / 50mg
Explore more special offers
Description
Description
CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.
Specifications
Specifications
CNX-2006
Gray
C26H27F4N7O2
DMSO : ≥ 52 mg/mL (95.32 mM)
545.53
99.68%
Solid
1375465-09-0
Research
50 mg
C=CC(NC1=CC=CC(NC2=NC(NC3=CC=C(NC4CN(CCF)C4)C=C3OC)=NC=C2C(F)(F)F)=C1)=O
545.53
Research
Cancer-Kinase/protease
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only