Learn More
Witeg™ Brown Glass Graduated Cylinders with Spout
Measure the volume of liquids in the laboratory with the Witeg™ Brown Glass Graduated Cylinders with Spout.
€ 12.70 - € 117.00
Specifications
Certifications/Compliance DIN EN ISO 4788
Material Borosilicate Glass
View More Specs
Products 8
Product Code Brand Capacity (Metric) Graduations Tolerance Metric Price Quantity & Availability  
Product Code Brand Capacity (Metric) Graduations Tolerance Metric Price Quantity & Availability  
10134444
View Documents
Witeg™
3 707 005
50 mL 2 mL ±1 mL
€ 12.70
Each
In Stock
to see stock.
Edge
Add to basket
10256644
View Documents
Witeg™
3 707 025
250 mL 5 mL ±2 mL
€ 55.35
Each
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
10452315
View Documents
Witeg™
3 707 050
500 mL 10 mL ±5 mL
€ 87.10
Each
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
10566014
View Documents
Witeg™
3 707 100
1000 mL 20 mL ±10 mL
€ 117.00
Each
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
10246644
View Documents
Witeg™
3 707 010
100 mL 2 mL ±1 mL
€ 17.95
Each
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
10298012
View Documents
Witeg™
3 705 010
100 mL 1 mL ±1 mL
€ 14.35
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
10023194
View Documents
Witeg™
3 705 100
1000 mL 10 mL ±10 mL
€ 61.80
Each
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
10358354
View Documents
Witeg™
3 715 010
100 mL 1 mL ±0.5 mL
€ 105.00
Pack of 10
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
Description
Description
  • Brown Glass
  • With Spout
  • Graduated
Specifications
Specifications
DIN EN ISO 4788
Borosilicate Glass