missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)naphthalene, 98%, Thermo Scientific Chemicals

€ 93.16 - € 475.00

Chemical Identifiers

CAS 7433-79-6
Molecular Formula C11H10S
Molecular Weight (g/mol) 174.261
MDL Number MFCD00051615
InChI Key SSKUUDUKJMIOKQ-UHFFFAOYSA-N
Synonym 2-methylthio naphthalene, 2-methylmercaptonaphthalene, 2-methylsulfanyl naphthalene, 2-methylthionaphthalene, methyl2-naphthylsulfide, methyl 2-naphthyl sulfide, naphth-2-yl methyl thioether, naphthalene,2-methylthio, methyl naphthalen-2-yl sulfane
PubChem CID 139022
IUPAC Name 2-methylsulfanylnaphthalene
SMILES CSC1=CC2=CC=CC=C2C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11480457
SDS View Documents
Thermo Scientific Alfa Aesar
B22368.03
1 g
€ 93.16
1g
Please to purchase this item. Need a web account? Register with us today!
11490457
SDS View Documents
Thermo Scientific Alfa Aesar
B22368.06
5 g
€ 361.80
5g
Please to purchase this item. Need a web account? Register with us today!
11400467
SDS View Documents
Thermo Scientific Alfa Aesar
B22368.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Methylthio)naphthalene, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.