missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)naphthalene, 98%

Product Code. 11400467
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11400467

Brand: Thermo Scientific Alfa Aesar B22368.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7433-79-6 | C11H10S | 174.261 g/mol

2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7433-79-6
Molecular Formula C11H10S
Molecular Weight (g/mol) 174.261
MDL Number MFCD00051615
InChI Key SSKUUDUKJMIOKQ-UHFFFAOYSA-N
Synonym 2-methylthio naphthalene, 2-methylmercaptonaphthalene, 2-methylsulfanyl naphthalene, 2-methylthionaphthalene, methyl2-naphthylsulfide, methyl 2-naphthyl sulfide, naphth-2-yl methyl thioether, naphthalene,2-methylthio, methyl naphthalen-2-yl sulfane
PubChem CID 139022
IUPAC Name 2-methylsulfanylnaphthalene
SMILES CSC1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 62°C to 63°C
Odor Stench
Quantity 25 g
Beilstein 1859409
Solubility Information Slightly soluble in water
Formula Weight 174.27
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)naphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.