Learn More
Thermo Scientific™ ZipChip™ Assay Kits
Reduce error and enjoy the convenience of prepackaged ZipChip™ Assay Kits, which are for use on the ZipChip Interface for analysis of biological samples by MS.
€ 1870.00 - € 5050.00
Specifications
For Use With (Equipment) ZipChip Interface for Mass Spectrometry
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
17881691
View Documents
Thermo Scientific™
00950-01-00500
Metabolites Assay Kit
€ 1870.00
1 set
17891691
View Documents
Thermo Scientific™
00950-01-00502
Intact Protein Assay Kit
€ 5050.00
1 set
Description
Description
Reduce error and enjoy the convenience of prepackaged ZipChip™ Assay Kits. These kits are for use with the ZipChip Interface, with or without the ZipChip Autosampler. Each ZipChip Assay Kit contains 5 bottles of background electrolyte and sample diluents, which is sufficient for 500 sample injections. A bottle can be directly loaded into ZipChip Autosampler for automated use and matching to a 96-well plate run. Different kits have been optimized for peptides, intact antibody, and metabolites analyses.
Three types of pre-packed ZipChip Assay Kits are customized for analyses of intact proteins, pepetides, and metabolites
  • Total of 500-sample capacity per kit
  • 5 individual bottles with 100 sample capacity each (designed to match to 96-well plate sampling) can be directly loaded into ZipChip autosampler
Specifications
Specifications
ZipChip Interface for Mass Spectrometry
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ ZipChip™ Assay Kits