Learn More
Thermo Scientific™ ZipChip™ Assay Kits
Reduce error and enjoy the convenience of prepackaged ZipChip™ Assay Kits, which are for use on the ZipChip Interface for analysis of biological samples by MS.
Brand:  Thermo Scientific™ 00950-01-00502
 View more versions of this product
Product Code. 17891691
  • € 5050.00 / 1 set
Explore more special offers
Description
Description
Reduce error and enjoy the convenience of prepackaged ZipChip™ Assay Kits. These kits are for use with the ZipChip Interface, with or without the ZipChip Autosampler. Each ZipChip Assay Kit contains 5 bottles of background electrolyte and sample diluents, which is sufficient for 500 sample injections. A bottle can be directly loaded into ZipChip Autosampler for automated use and matching to a 96-well plate run. Different kits have been optimized for peptides, intact antibody, and metabolites analyses.
Three types of pre-packed ZipChip Assay Kits are customized for analyses of intact proteins, pepetides, and metabolites
  • Total of 500-sample capacity per kit
  • 5 individual bottles with 100 sample capacity each (designed to match to 96-well plate sampling) can be directly loaded into ZipChip autosampler
Specifications
Specifications
Intact Protein Assay Kit
Intact Protein Assay Kit
ZipChip Interface for Mass Spectrometry
Background electrolyte and sample diluents optimized for intact protien analysis
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ ZipChip™ Assay Kits >