Learn More
Tocris Bioscience™ Wnt-C59
Highly potent PORCN inhibitor
Brand:  Tocris Bioscience™ 5148/50
 View more versions of this product
Product Code. 18735708
  • € 1470.00 / 50mg
Explore more special offers
Description
Description
Wnt-C59 is a highly potent inhibitor of MBOAT (membrane-bound O-acyltranferase) family member Porcupine (PORCN) (IC50 = 74 pM) that mediates WNT palmitoylation and secretion. Wnt-C59 potently inhibits the processing of both canonical (1, 2, 3a, 6, 7b, 8a, 9a, 9b, 10) and non-canonical (4, 5a, 11, 16) Wnt subtypes. Wnt-C59 blocks progression of mammary tumors in MMTV-WNT1 transgenic mice and downregulates Wnt/β-catenin target genes. Wnt-C59 treated tumors show a decrease in β-catenincatenin, CyclinD1 and c-Myc. Wnt-C59 induces cardiomyocyte differentiation from human iPSCs following culture with CHIR 99021 (Cat. No. 4423). Wnt-C59 efficiently induces neural differentiation of CTIP2+/COUP-TF1- cells from PSCs in culture. When grafted into the cortex of adult mice, develop abundant axonal fiber extension toward the spinal cord. Cell permeable and orally bioavailable.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
>99%
Wnt-C59
Wnt-C59
50 mg
379.45