Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes feature a susceptibility matched bottom and plunger to effectively reduce the boundary gradients at the edges of the Rf coil reducing the required sample volume by two thirds.
€ 312.00 - € 357.00
Specifications
Product Type Tube Set
View More Specs
Products 6
Product Code Brand Diameter (Metric) Outer For Use With (Equipment) Model Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer For Use With (Equipment) Model Price Quantity & Availability  
15388588
View Documents Promotion Details
Bel-Art™ SP Scienceware™
66000-0027
3 mm Universal, All Spectometers BMS-003
€ 319.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
15398588
View Documents Promotion Details
Bel-Art™ SP Scienceware™
66000-0028
5 mm Bruker Spectometers BMS-005B
€ 313.00
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
15338598
View Documents Promotion Details
Bel-Art™ SP Scienceware™
66000-0030
3 mm Universal, All Spectometers CMS-003
€ 312.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
15358598
View Documents Promotion Details
Bel-Art™ SP Scienceware™
66000-0031
5 mm Bruker Spectometers CMS-005B
€ 319.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
15368598
View Documents Promotion Details
Bel-Art™ SP Scienceware™
66000-0032
3 mm Universal, All Spectometers DMS-003
€ 357.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
15308608
View Documents Promotion Details
Bel-Art™ SP Scienceware™
66000-0036
3 mm Universal, All Spectometers MMS-003
€ 357.00
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
Description
Description
  • Select tube based on solvent used: CDCL3, CD3OD, D2O, DMSO-d6
  • Available for Bruker or Universal
Specifications
Specifications
Tube Set