Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes feature a susceptibility matched bottom and plunger to effectively reduce the boundary gradients at the edges of the Rf coil reducing the required sample volume by two thirds.
Brand: Bel-Art™ SP Scienceware™ 66000-0030
This item is not returnable.
View return policy
Description
- Select tube based on solvent used: CDCL3, CD3OD, D2O, DMSO-d6
- Available for Bruker or Universal
Specifications
Universal, All Spectometers | |
Tube Set | |
Chloroform-d Matched |
10 mm (Bottom) | |
3 mm | |
CMS-003 |