Learn More
Gosselin™ White Straight Specimen Containers with Red Cap and Spoon
Corning™ Gosselin™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
Brand: Gosselin™ TP35C-025
330.49 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Clear and break-resistant polypropylene (PP)
- Available in different sizes and quantities
- Color-coded screw caps
Specifications
Specimen Container | |
Polypropylene | |
Sterile | |
Red | |
White | |
No Label | |
70/Bag |
Yes | |
Straight | |
60 mL | |
Screw Cap | |
Cap with white spoon | |
700/Cs. |
Product Content Correction
Product Title
Gosselin™ White Straight Specimen Containers with Red Cap and Spoon > 60mL