Learn More
Testo™ Waterproof Surface Probe
Brand: Testo™ 0609 1973
This item is not returnable.
View return policy
Specifications
Probe, Surface | |
EN 13485 and HACCP-compliant | |
40 sec. | |
-50°C to +400°C |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Testo™ Waterproof Surface Probe > Probe Diameter: 5mm