Learn More
Tocris Bioscience™ VU 0463271
Potent and selective KCC2 inhibitor
Brand:  Tocris Bioscience™ 4719/50
 View more versions of this product
Product Code. 18761389
  • € 1005.37 / 50mg
Explore more special offers
Description
Description
VU 0463271 is a potent and selective inhibitor of the neuronal K-Cl cotransporter, KCC2 (IC50 = 61 nM); displays >100-fold selectivity versus the Na-K-2Cl cotransporter 1 (NKCC1) and no activity against a panel of 68 GPCRs, ion channels and transporters.
Specifications
Specifications
N-Cyclopropyl-N-(4-methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]acetamide
50 mg
C19H18N4OS2
>98%
1391737-01-1
KCC2 Inhibitors
382.5