missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 0463271
Potent and selective KCC2 inhibitor
€ 254.85 - € 1005.37
Specifications
Chemical Name or Material | N-Cyclopropyl-N-(4-methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]acetamide |
---|---|
CAS | 1391737-01-1 |
Target | KCC2 Inhibitors |
Molecular Formula | C19H18N4OS2 |
Formula Weight | 382.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774049
|
Tocris Bioscience™
4719/10 |
10 mg |
€ 254.85
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18761389
|
Tocris Bioscience™
4719/50 |
50 mg |
€ 1005.37
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective inhibitor of the neuronal K-Cl cotransporter, KCC2 (IC50 = 61 nM); displays >100-fold selectivity versus the Na-K-2Cl cotransporter 1 (NKCC1) and no activity against a panel of 68 GPCRs, ion channels and transporters.Specifications
N-Cyclopropyl-N-(4-methyl-2-thiazolyl)-2-[(6-phenyl-3-pyridazinyl)thio]acetamide | |
KCC2 Inhibitors | |
382.5 |
1391737-01-1 | |
C19H18N4OS2 | |
>98% |
Safety and Handling
Recommended Storage : Store at 4°C