Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Amber Graduation
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Amber Graduation is ideal for measuring and transferring various quantities of liquid.
Brand:  Poulten & Graf GmbH 1 11437404
 View more versions of this product
Product Code. 15962100
  • € 19.20 / Pack of 5
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Description
  • Total delivery
  • Amber graduation
  • With ring marks
  • Comformity approved
Specifications
Specifications
EN ISO 835
Graduated pipette
Non-sterile
Amber
0.05 mL
Graduated Pipet
AS
Soda Lime Glass
Yes
Amber
10 mL
Reusable