Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Amber Graduation
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Amber Graduation is ideal for measuring and transferring various quantities of liquid.
Brand: Poulten & Graf GmbH 1 11427404
This item is not returnable.
View return policy
Description
- Total delivery
- Amber graduation
- With ring marks
- Comformity approved
Specifications
EN ISO 835 | |
Graduated pipette | |
Non-sterile | |
Amber | |
2 mL | |
Reusable |
AS | |
Soda Lime Glass | |
Yes | |
0.01 mL | |
Graduated Pipet |