Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Amber Graduation
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Amber Graduation is ideal for measuring and transferring various quantities of liquid.
Brand:  Poulten & Graf GmbH 1 11447404
 View more versions of this product
Product Code. 10688592
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Total delivery
  • Amber graduation
  • With ring marks
  • Comformity approved
Specifications
Specifications
EN ISO 835
Graduated pipette
Non-sterile
Amber
50 mL
Reusable
AS
Soda Lime Glass
Yes
0.20 mL
Graduated Pipet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers