Learn More
HUBER Variostat™ Bath
Variostat™ Bath insulated stainless steel tanks are the ideal choice to incubate samples and keep them at a constant temperature.
Brand:  HUBER 6052
 View more versions of this product
Product Code. 10013764
  • € 1155.00 / Each
Estimated Shipment: 19-06-2025
to see stock.
Explore more special offers
Description
  • Insulated, stainless steel tanks
  • Drain port
  • Bath cover sold separately
Specifications
Specifications
5.5 L
210 mm
230 V 50 Hz
50 Hz
30°C to 150°C
205 mm
230 V
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
HUBER Variostat™ Bath > Variostat™ Bath; Capacity: 5.5 L