Learn More
Heinz Herenz Dissection Sets
Dissection sets with wooden case.
Brand: Heinz Herenz 1110401
This item is not returnable.
View return policy
Description
- Dissection sets
- Strong wooden case: 19 x 9 x 3.5 cm
- Contents: 1 scalpel handle No. 4 with 2 blades, 1 scalpel, 1 scalpel handle No. 3 with 2 blades, 1 forceps, 1 scissors (pointed), 2 dissection needles (1x pointed, 1x with blade)
Specifications
Dissection Set | |
Reusable | |
Non-sterile |
Product Content Correction
Product Title
Heinz Herenz Dissection Sets > With Wooden Case