Learn More
BINDER™ Pump Chamber with VAP 2 Vacuum Pump
Pump Chamber with VAP 2 Vacuum Pump - 230 V / 50-60 Hz option model, pump chamber for vacuum pumps, with switchable power socket. Chemical membrane pump with a nominal air flow of 4.0 m³/hour, final pressure 1.8 mbar
€ 7550.00 - € 7760.00
Specifications
Max. Vacuum | 1011.45 mbar below atmospheric pressure |
---|---|
Electrical Requirements | 230 V, 50/60 Hz |
Description | Pump chamber with VAP 2 vacuum pump - 230 V / 50-60 Hz option model, Pump chamber for vacuum pumps, with switchable power socket. Chemical membrane pump with a nominal air flow of 4.0 m3/hour, final pressure 1,8 Mbar |
Flow Rate | 4.0 m3/hr. |
Frequency | 50/60 Hz |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
16676463
|
BINDER™
8012-1964 |
BINDER Chambers VD 23 |
€ 7550.00
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
|||||
16664392
|
BINDER™
8012-1965 |
BINDER Chambers VD 56 |
€ 7685.00
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
|||||
16686463
|
BINDER™
8012-1966 |
BINDER Chambers VD 115 |
€ 7760.00
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
Description
Pump chamber:
- Made of powder-coated sheet metal, with large viewing window
Pump:
- Nominal air flow: 4.0 m³/hr.
- End vacuum: 1.8 mbar
- Pump membrane made of highly resistant PEEK plastic
- Noise level < 50 dB(A)
Specifications
1011.45 mbar below atmospheric pressure | |
Pump chamber with VAP 2 vacuum pump - 230 V / 50-60 Hz option model, Pump chamber for vacuum pumps, with switchable power socket. Chemical membrane pump with a nominal air flow of 4.0 m3/hour, final pressure 1,8 Mbar | |
50/60 Hz | |
IP55 | |
230 V |
230 V, 50/60 Hz | |
4.0 m3/hr. | |
Vacuum Pump | |
2 Mbar |