Learn More
Superfos™ UniPak™ Polypropylene Bucket
Provides flexible packaging. Superfos™ UniPak™ Polypropylene Buckets match each lid diameter so only one versatile type of lid is needed for a range of sizes. Tamper resistant.
Brand: Superfos™ 5030-WHITE
Description
- Protective collar
- Withstands hot filling and blast freezing
- Available in round, rectangular, oval and square shapes
- Broad range of standard sizes
- Liquid tight, snap-on lid
- Same lid fits several volumes
- Excellent decoration options with In-Mould Labelling
- Visible deterrent to tampering
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.