missing translation for 'onlineSavingsMsg'
Learn More

SCA Hygiene Products™ Tork Virgin Fibre Wiping Paper

Brand:  SCA Hygiene Products™ 130072

 View more versions of this product

Product Code. 10033364

  • € 85.00 / 2 rolls
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Wiping Paper
235 mm
ISO 9001:2000, ISO 14001 and EMAS
Single-use
Roll
2 Rolls
170 m
Virgin Fiber
Blue
170 m x 235 mm
Tork Floor Stand, Tork Performance Floor Stand Mobile White, Tork Wall Stand
2-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers