Learn More
Topiramate, MedChemExpress
Product Code. 16458116
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16458116
Brand: Medchem Express HYB0122
MedChemExpress Topiramate (McN 4853) is a broad-spectrum antiepileptic agent. Topiramate is a GluR5 receptor antagonist. Topiramate produces its antiepileptic effects through enhancement of GABAergic activity, inhibition of kainate/AMPA receptors, inhibition of voltage-sensitive sodium and calcium channels, increases in potassium conductance, and inhibition of carbonic anhydrase.
Topiramate (McN 4853) is a broad-spectrum antiepileptic agent. Topiramate is a GluR5 receptor antagonist. Topiramate produces its antiepileptic effects through enhancement of GABAergic activity, inhibition of kainate/AMPA receptors, inhibition of voltage-sensitive sodium and calcium channels, increases in potassium conductance, and inhibition of carbonic anhydrase.
Specifications
Chemical Name or Material Topiramate
CAS 97240-79-4
Color White
Purity Grade Notes Research
Molecular Formula C12H21NO8S
Quantity 100 mg
Synonym McN 4853 RWJ 17021
Solubility Information DMSO : ≥ 100 mg/mL (294.67 mM) ∣H2O : 4 mg/mL (11.79 mM; Need ultrasonic)
SMILES NS(OC[C@]1(OC(C)(C)O2)[C@@H]2[C@H](OC(C)(C)O3)[C@H]3CO1)(=O)=O
Molecular Weight (g/mol) 339.36
Formula Weight 339.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only