Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key, threaded, hollow
For securing the hollow DURAN™ tap plug in the ground DURAN™ single way stopcock.
€ 87.70 - € 389.00
Specifications
Material | Borosilicate Glass (3.3), Polypropylene |
---|---|
Autoclavable | Autoclavable |
No. per Pack | 100/Pk. |
For Use With (Application) | For securing the hollow DURAN™ tap plug in the ground DURAN™ one-way tap |
Product Code | Brand | Closure Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | ||||||
15844528
|
DWK Life Sciences
286041409 |
12.5 NS |
€ 87.70
Pack of 100 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
||||||
15894528
|
DWK Life Sciences
286046004 |
21.5 NS |
€ 156.00
Pack of 100 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
||||||
15804538
|
DWK Life Sciences
286046107 |
24, 29.2 NS |
€ 389.00
Pack of 100 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
||||||
15874528
|
DWK Life Sciences
286044209 |
18.8 NS |
€ 109.00
Pack of 100 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
||||||
12848041
|
DWK Life Sciences
286042808 |
14.5 NS |
€ 107.00
Pack of 100 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
||||||
Description
The 3-part tap safety device holds and secures the hollow DURAN™ tap plug in the tap sleeve.
- 3-part, for hollow tap plugs
- Three part retaining device holds and secures the tap plug in the tap sleeve
Specifications
Borosilicate Glass (3.3), Polypropylene | |
Autoclavable | |
100/Pk. | |
For securing the hollow DURAN™ tap plug in the ground DURAN™ one-way tap |
Product Title
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key, threaded, hollow