Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key, threaded, hollow
For securing the hollow DURAN™ tap plug in the ground DURAN™ single way stopcock.
Brand:  DWK Life Sciences 286044209
 View more versions of this product
Product Code. 15874528
  • € 109.00 / Pack of 100
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Description
The 3-part tap safety device holds and secures the hollow DURAN™ tap plug in the tap sleeve.
  • 3-part, for hollow tap plugs
  • Three part retaining device holds and secures the tap plug in the tap sleeve
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass (3.3), Polypropylene
18.8 NS
For securing the hollow DURAN™ tap plug in the ground DURAN™ one-way tap
Autoclavable
100/Pk.