Learn More
SCAT™ PTFE Thread Adapters
SCAT™ Safety Caps and SCAT™ Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
€ 38.40 - € 227.82
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
10120830
|
SCAT™
107992 |
GL38 (f) to GL45 (m) |
€ 88.15
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
10374900
|
SCAT™
107993 |
GL32 (f) to GL45 (m) |
€ 83.35
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
10318729
|
SCAT™
107017 |
GL45 (m) to 2 in. Tri-sure (m) |
€ 116.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
10829840
|
SCAT™
107016 |
GL45 (m) to R2 in. BSP/G2 in. (m) |
€ 116.00
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
10840411
|
SCAT™
117094 |
GL45 (f) to S55 (m) |
€ 83.35
Each |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
10860391
|
SCAT™
107509 |
GL45 (m) to NS29/32 mm (m) |
€ 138.00
Each |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
10384420
|
SCAT™
107991 |
GL/S40 (f) to GL45 (m) |
€ 83.35
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
10705802
|
SCAT™
117091 |
GL/S40 (f) to S55 (m) |
€ 83.35
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
10881051
|
SCAT™
117095 |
S51 (f) to S55 (m) |
€ 83.35
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
10584448
|
SCAT™
107632 |
UNF 1/4 in. 28 G (f) to GL14 (m) |
€ 38.40
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
10130200
|
SCAT™
117030 |
GPI 38-23 (f) to GL45 (m) |
€ 83.35
Each |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
10671211
|
SCAT™
160146 |
ND9 |
€ 44.80
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||||
17917270
|
SCAT™
108325 |
S60/61 (m) to B83 (f) |
€ 227.82
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SCAT™ thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polytetrafluoroethylene (PTFE) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifications
PTFE | |
Thread Adapter |